SY3H3D67FCCC Synthonix Corporation

5-Methoxy-2-nitrophenol

  • MDL number No MDL found
  • Cas Number : 704-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45859.html

Safety Information

MSDS https://synthonix.com/45859.html
Availability details for
Quantity
  • Availability
  • Add to Cart