SY3H3D67FCCF Synthonix Corporation

Methyl 5-bromo-2-methylthiophene-3-carboxylate

  • Cas Number : 1259396-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45909.html

Safety Information

MSDS https://synthonix.com/45909.html
Availability details for
Quantity
  • Availability
  • Add to Cart