SY3H3D67FCDF Synthonix Corporation

1-(2-Methoxypyrimidin-5-yl)ethanone

  • Cas Number : 1056174-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46010.html

Safety Information

MSDS https://synthonix.com/46010.html
Availability details for
Quantity
  • Availability
  • Add to Cart