SY3H3D67FCE0 Synthonix Corporation

Methyl 5-methylpyrazine-2-carboxylate

  • Cas Number : 41110-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46011.html

Safety Information

MSDS https://synthonix.com/46011.html
Availability details for
Quantity
  • Availability
  • Add to Cart