SY3H3D67FCE1 Synthonix Corporation

Methyl 6-methylpyrazine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 41110-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46012.html

Safety Information

MSDS https://synthonix.com/46012.html
Availability details for
Quantity
  • Availability
  • Add to Cart