SY3H3D67FCE7 Synthonix Corporation

6-Methoxy-2-methyl-3-nitropyridine

  • Cas Number : 5467-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46033.html

Safety Information

MSDS https://synthonix.com/46033.html
Availability details for
Quantity
  • Availability
  • Add to Cart