SY3H3D67FCE8 Synthonix Corporation

Methyl 6-amino-5-hydroxynicotinate

  • Cas Number : 1260663-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46034.html

Safety Information

MSDS https://synthonix.com/46034.html
Availability details for
Quantity
  • Availability
  • Add to Cart