SY3H3D67FCE9 Synthonix Corporation

Methyl 6-methoxy-2-pyrazinecarboxylate

  • MDL number No MDL found
  • Cas Number : 23813-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46038.html

Safety Information

MSDS https://synthonix.com/46038.html
Availability details for
Quantity
  • Availability
  • Add to Cart