SY3H3D67FCEB Synthonix Corporation

Methyl 2,4-dihydroxy-6-methylpyrimidine-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 869891-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46053.html

Safety Information

MSDS https://synthonix.com/46053.html
Availability details for
Quantity
  • Availability
  • Add to Cart