SY3H3D67FCEE Synthonix Corporation

2-Methyl-5-nitrobenzenesulfonamide

  • MDL number MFCD00115503
  • Cas Number : 6269-91-6
  • Molecular Weight : 216.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Off-white to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/46056.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46056.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart