SY3H3D67FCF0 Synthonix Corporation

2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine

  • Cas Number : 36267-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46059.html

Safety Information

MSDS https://synthonix.com/46059.html
Availability details for
Quantity
  • Availability
  • Add to Cart