SY3H3D67FCF1 Synthonix Corporation

Methyl 6-bromo-3-(methylamino)pyrazine-2-carboxylate

  • Cas Number : 13457-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46064.html

Safety Information

MSDS https://synthonix.com/46064.html
Availability details for
Quantity
  • Availability
  • Add to Cart