SY3H3D67FCF4 Synthonix Corporation

(2-Methoxy-5-nitrophenyl)boronic acid

  • Cas Number : 677746-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46149.html

Safety Information

MSDS https://synthonix.com/46149.html
Availability details for
Quantity
  • Availability
  • Add to Cart