SY3H3D67FCF8 Synthonix Corporation

Methyl 2-methylthiophene-3-carboxylate

  • Cas Number : 53562-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46248.html

Safety Information

MSDS https://synthonix.com/46248.html
Availability details for
Quantity
  • Availability
  • Add to Cart