SY3H3D67FCF9 Synthonix Corporation

Methyl 3-methylthiophene-2-carboxylate

  • MDL number MFCD00234332
  • Cas Number : 81452-54-2
  • Molecular Weight : 156.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/46249.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46249.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart