SY3H3D67FD07 Synthonix Corporation

N1-Methyl-4-nitrobenzene-1,2-diamine

  • MDL number MFCD00156607
  • Cas Number : 41939-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46347.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46347.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart