SY3H3D67FD0B Synthonix Corporation

2-Methoxy-3-methylpyridine

  • MDL number No MDL found
  • Cas Number : 19230-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46378.html

Safety Information

MSDS https://synthonix.com/46378.html
Availability details for
Quantity
  • Availability
  • Add to Cart