SY3H3D67FD0F Synthonix Corporation

(4-Methylpyridin-2-yl)methanol

  • MDL number No MDL found
  • Cas Number : 42508-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46387.html

Safety Information

MSDS https://synthonix.com/46387.html
Availability details for
Quantity
  • Availability
  • Add to Cart