SY3H3D67FD14 Synthonix Corporation

Methyl 5-methyl-1H-pyrrole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1194-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/46412.html

Safety Information

MSDS https://synthonix.com/46412.html
Availability details for
Quantity
  • Availability
  • Add to Cart