SY3H3D67FD22 Synthonix Corporation

4-Methoxybenzene-1,2-diamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 106658-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46463.html

Safety Information

MSDS https://synthonix.com/46463.html
Availability details for
Quantity
  • Availability
  • Add to Cart