SY3H3D67FD23 Synthonix Corporation

(3-Methoxypyridin-2-yl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1588441-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46464.html

Safety Information

MSDS https://synthonix.com/46464.html
Availability details for
Quantity
  • Availability
  • Add to Cart