SY3H3D67FD27 Synthonix Corporation

2-Methyl-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine hydrochloride

  • Cas Number : 1159011-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46472.html

Safety Information

MSDS https://synthonix.com/46472.html
Availability details for
Quantity
  • Availability
  • Add to Cart