SY3H3D67FD28 Synthonix Corporation

2-Morpholinothiazol-4-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 170492-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46474.html

Safety Information

MSDS https://synthonix.com/46474.html
Availability details for
Quantity
  • Availability
  • Add to Cart