SY3H3D67FD2B Synthonix Corporation

1-Methylpiperidine-2-carboxylic acid hydrochloride

  • Cas Number : 25271-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46486.html

Safety Information

MSDS https://synthonix.com/46486.html
Availability details for
Quantity
  • Availability
  • Add to Cart