SY3H3D67FD32 Synthonix Corporation

(S)-1-(3-Methylpiperazin-1-yl)ethanone hydrochloride

  • MDL number No MDL found
  • Cas Number : 1638137-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46501.html

Safety Information

MSDS https://synthonix.com/46501.html
Availability details for
Quantity
  • Availability
  • Add to Cart