SY3H3D67FD3C Synthonix Corporation

2-Methyl-4-(trifluoromethyl)pyridine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1416354-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46531.html

Safety Information

MSDS https://synthonix.com/46531.html
Availability details for
Quantity
  • Availability
  • Add to Cart