SY3H3D67FD3F Synthonix Corporation

4-Methylpicolinimidamide hydrochloride

  • MDL number No MDL found
  • Cas Number : 1179360-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46572.html

Safety Information

MSDS https://synthonix.com/46572.html
Availability details for
Quantity
  • Availability
  • Add to Cart