SY3H3D67FD40 Synthonix Corporation

6-Methoxynicotinimidamide hydrochloride

  • MDL number No MDL found
  • Cas Number : 201937-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/46573.html

Safety Information

MSDS https://synthonix.com/46573.html
Availability details for
Quantity
  • Availability
  • Add to Cart