SY3H3D67FD41 Synthonix Corporation

Methyl 4-(aminomethyl)thiophene-2-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 916214-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46579.html

Safety Information

MSDS https://synthonix.com/46579.html
Availability details for
Quantity
  • Availability
  • Add to Cart