SY3H3D67FD46 Synthonix Corporation

(6-Methoxypyridin-2-yl)methanamine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 2172852-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46592.html

Safety Information

MSDS https://synthonix.com/46592.html
Availability details for
Quantity
  • Availability
  • Add to Cart