SY3H3D67FD48 Synthonix Corporation

5-Methyl-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 92223-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46597.html

Safety Information

MSDS https://synthonix.com/46597.html
Availability details for
Quantity
  • Availability
  • Add to Cart