SY3H3D67FD49 Synthonix Corporation

1-Methyl-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine dihydrochloride

  • Cas Number : 1228878-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46598.html

Safety Information

MSDS https://synthonix.com/46598.html
Availability details for
Quantity
  • Availability
  • Add to Cart