SY3H3D67FD4A Synthonix Corporation

2-(((4-Methyl-1H-imidazol-5-yl)methyl)thio)ethanamine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 38603-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46600.html

Safety Information

MSDS https://synthonix.com/46600.html
Availability details for
Quantity
  • Availability
  • Add to Cart