SY3H3D67FD4D Synthonix Corporation

3-Methylbenzoic hydrazide

  • MDL number No MDL found
  • Cas Number : 13050-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46653.html

Safety Information

MSDS https://synthonix.com/46653.html
Availability details for
Quantity
  • Availability
  • Add to Cart