SY3H3D67FD4F Synthonix Corporation

4-Methyl-3,4-dihydropyrrolo[1,2-a]pyrazin-1(2H)-one

  • MDL number No MDL found
  • Cas Number : 1243415-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46658.html

Safety Information

MSDS https://synthonix.com/46658.html
Availability details for
Quantity
  • Availability
  • Add to Cart