SY3H3D67FD53 Synthonix Corporation

N-Methyl-1-(4-nitrophenyl)methanamine

  • MDL number No MDL found
  • Cas Number : 19499-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46683.html

Safety Information

MSDS https://synthonix.com/46683.html
Availability details for
Quantity
  • Availability
  • Add to Cart