SY3H3D67FD55 Synthonix Corporation

4-Methoxy-5-methyl-2-nitroaniline

  • MDL number No MDL found
  • Cas Number : 55730-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46691.html

Safety Information

MSDS https://synthonix.com/46691.html
Availability details for
Quantity
  • Availability
  • Add to Cart