SY3H3D67FD63 Synthonix Corporation

(3-(Methoxycarbamoyl)phenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 850567-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46783.html

Safety Information

MSDS https://synthonix.com/46783.html
Availability details for
Quantity
  • Availability
  • Add to Cart