SY3H3D67FD68 Synthonix Corporation

(3-(Methylsulfonyl)phenyl)methanol

  • MDL number No MDL found
  • Cas Number : 220798-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/46853.html

Safety Information

MSDS https://synthonix.com/46853.html
Availability details for
Quantity
  • Availability
  • Add to Cart