SY3H3D67FD69 Synthonix Corporation

Methyl 5-(2-hydroxyethyl)thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 160744-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/46854.html

Safety Information

MSDS https://synthonix.com/46854.html
Availability details for
Quantity
  • Availability
  • Add to Cart