SY3H3D67FD70 Synthonix Corporation

4-(Methoxymethyl)aniline

  • MDL number No MDL found
  • Cas Number : 80936-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46895.html

Safety Information

MSDS https://synthonix.com/46895.html
Availability details for
Quantity
  • Availability
  • Add to Cart