SY3H3D67FD79 Synthonix Corporation

4-Methyl-3-(methylamino)phenol

  • MDL number No MDL found
  • Cas Number : 6265-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46910.html

Safety Information

MSDS https://synthonix.com/46910.html
Availability details for
Quantity
  • Availability
  • Add to Cart