SY3H3D67FD7A Synthonix Corporation

(4-Methoxy-3-methylpyridin-2-yl)methanol

  • MDL number No MDL found
  • Cas Number : 86604-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46930.html

Safety Information

MSDS https://synthonix.com/46930.html
Availability details for
Quantity
  • Availability
  • Add to Cart