SY3H3D67FD7B Synthonix Corporation

Methyl 2-(1-methyl-1H-pyrrol-2-yl)acetate

  • Cas Number : 51856-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46931.html

Safety Information

MSDS https://synthonix.com/46931.html
Availability details for
Quantity
  • Availability
  • Add to Cart