SY3H3D67FD7C Synthonix Corporation

Methyl 2-(2,5-dimethylthiazol-4-yl)acetate

  • MDL number No MDL found
  • Cas Number : 306937-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46934.html

Safety Information

MSDS https://synthonix.com/46934.html
Availability details for
Quantity
  • Availability
  • Add to Cart