SY3H3D67FD82 Synthonix Corporation

(5-Methoxy-2-methylphenyl)boronic acid

  • MDL number MFCD03425973
  • Cas Number : 617689-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46950.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46950.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart