SY3H3D67FD85 Synthonix Corporation

2-Methyl-6-propoxypyrazine

  • Cas Number : 67845-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46976.html

Safety Information

MSDS https://synthonix.com/46976.html
Availability details for
Quantity
  • Availability
  • Add to Cart