SY3H3D67FD88 Synthonix Corporation

Methyl 2-amino-5-propylthiazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 649736-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46997.html

Safety Information

MSDS https://synthonix.com/46997.html
Availability details for
Quantity
  • Availability
  • Add to Cart