SY3H3D67FD8B Synthonix Corporation

Methyl 3-oxocyclohexanecarboxylate

  • Cas Number : 13148-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47023.html

Safety Information

MSDS https://synthonix.com/47023.html
Availability details for
Quantity
  • Availability
  • Add to Cart