SY3H3D67FD8C Synthonix Corporation

Methyl 4-oxocyclohexanecarboxylate

  • MDL number No MDL found
  • Cas Number : 6297-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/47024.html

Safety Information

MSDS https://synthonix.com/47024.html
Availability details for
Quantity
  • Availability
  • Add to Cart